2,5-Dichloronicotinoyl chloride

Lieferant: Apollo Scientific
Gefahr

OR29880-100MG
APOSOR29880-100MGEA 21.9 EUR
APOSOR29880-100MG
2,5-Dichloronicotinoyl chloride
2,5-Dichloronicotinoyl chloride
Formel: C₆H₂Cl₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00210149
CAS-Nummer: 78686-87-0
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR