3-Amino-2,5-dichlorpyridin 96%
Lieferant: Apollo Scientific
|
Achtung
|
OR29881-5G
APOSOR29881-5GEA
21.9
EUR
APOSOR29881-5G
3-Amino-2,5-dichlorpyridin 96%
3-Amino-2,5-dichlorpyridin
Formel:
C₅H₄Cl₂N₂ Molecular Weight: 163,01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00204157 CAS-Nummer: 78607-32-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...