2-Chlorisonicotinamid

Lieferant: Apollo Scientific
Achtung

OR29886-1G
APOSOR29886-1GEA 21.9 EUR
APOSOR29886-1G
2-Chlorisonicotinamid
2-Chlorisonicotinamid
Formel: C₆H₅ClN₂O
Molecular Weight: 156.57 g/mol
Schmelzpunkt: 201…203 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00221401
CAS-Nummer: 100859-84-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR