Ethyl {[5-(2,6-dichloropyridin-4-yl)-4-methyl-4H-1,2,4-triazol-3-yl]thio}acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl {[5-(2,6-dichloropyridin-4-yl)-4-methyl-4H-1,2,4-triazol-3-yl]sulphanyl}acetate, Ethyl 2-{[5-(2,6-dichloropyridin-4-yl)-4-methyl-4H-1,2,4-triazol-3-yl]sulphanyl}ethanoate

OR29906-50MG
APOSOR29906-50MGEA 43.5 EUR
APOSOR29906-50MG
Ethyl {[5-(2,6-dichloropyridin-4-yl)-4-methyl-4H-1,2,4-triazol-3-yl]thio}acetate
Ethyl {[5-(2,6-dichloropyridin-4-yl)-4-methyl-4H-1,2,4-triazol-3-yl]thio}acetate
Formel: C₁₂H₁₂Cl₂N₄O₂S
Molecular Weight: 347.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00833056
CAS-Nummer: 680216-57-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR