2-Chloro-4-{3-[(4-methylphenoxy)methyl]-1,2,4-oxadiazol-5-yl}pyridine

Lieferant: Apollo Scientific
Achtung

OR29921-100MG
APOSOR29921-100MGEA 279 EUR
APOSOR29921-100MG
2-Chloro-4-{3-[(4-methylphenoxy)methyl]-1,2,4-oxadiazol-5-yl}pyridine
2-Chloro-4-{3-[(4-methylphenoxy)methyl]-1,2,4-oxadiazol-5-yl}pyridine
Formel: C₁₅H₁₂ClN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00833093
CAS-Nummer: 680216-74-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR