2-Chlor-6-methyl-3-nitropyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Chlor-5-nitro-2-picolin, 6-Chlor-2-methyl-5-nitropyridin, 2-Chlor-3-nitro-6-picolin
OR2994-1G
APOSOR2994-1GEA
21.9
EUR
APOSOR2994-1G
2-Chlor-6-methyl-3-nitropyridin
2-Chloro-6-methyl-3-nitropyridine
Formel:
C₆H₅ClN₂O₂ Schmelzpunkt: 70…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03085820 CAS-Nummer: 56057-19-3 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...