3-Oxo-3-pyridin-4-ylpropanenitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Oxo-3-(pyridin-4-yl)propanenitrile

OR2995-1G
APOSOR2995-1GEA 184 EUR
APOSOR2995-1G
3-Oxo-3-pyridin-4-ylpropanenitrile
3-Oxo-3-pyridin-4-ylpropanenitrile
Formel: C₈H₆N₂O
Molecular Weight: 146,15 g/mol
Schmelzpunkt: 95…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06656807
CAS-Nummer: 23821-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR