4-Methoxy-3-nitrobenzamid
Lieferant: Apollo Scientific
OR29952-1G
APOSOR29952-1GEA
84.1
EUR
APOSOR29952-1G
4-Methoxy-3-nitrobenzamid
4-Methoxy-3-nitrobenzamid
Formel:
C₈H₈N₂O₃ Molecular Weight: 180,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017136 CAS-Nummer: 10397-58-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...