Thiophene-2-carboxamidine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR2996-1G OR2996-5G
APOSOR2996-1GEA 0 EUR
APOSOR2996-1G APOSOR2996-5G
Thiophene-2-carboxamidine hydrochloride
Thiophene-2-carboxamidine hydrochloride
Formel: C₅H₇ClN₂S
Schmelzpunkt: 173…177 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00173786
CAS-Nummer: 54610-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR