3-Amino-5-brom-2-chlorpyridin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Brom-2-chlor-3-pyridylamin
OR29961-5G
APOSOR29961-5GEA
21.9
EUR
APOSOR29961-5G
3-Amino-5-brom-2-chlorpyridin 98%
3-Amino-5-bromo-2-chloropyridine
Formel:
C₅H₄BrClN₂ Molecular Weight: 207,46 g/mol Schmelzpunkt: 120…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682092 CAS-Nummer: 588729-99-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...