4,6-Dichloro-2-(methylthio)pyrimidine-5-carbonitrile

Lieferant: Apollo Scientific

OR29981-5G
APOSOR29981-5GEA 94.4 EUR
APOSOR29981-5G
4,6-Dichloro-2-(methylthio)pyrimidine-5-carbonitrile
4,6-Dichloro-2-(methylthio)pyrimidine-5-carbonitrile
Formel: C₆H₃Cl₂N₃S
Molecular Weight: 220,0800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00474490
CAS-Nummer: 33097-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR