1-Chloroisoquinoline-5-boronic acid

Lieferant: Apollo Scientific
Achtung

OR300001-250MG
APOSOR300001-250MGEA 0 EUR
APOSOR300001-250MG
1-Chloroisoquinoline-5-boronic acid
1-Chloroisoquinoline-5-boronic acid
Formel: C₉H₇BClNO₂
Lagertemperatur: Kühlschrank
MDL: MFCD18257026
CAS-Nummer: 2096338-36-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR