3-Chloroisoquinoline-6-boronic acid

Lieferant: Apollo Scientific
Achtung

OR300005-250MG
APOSOR300005-250MGEA 140 EUR
APOSOR300005-250MG
3-Chloroisoquinoline-6-boronic acid
3-Chloroisoquinoline-6-boronic acid
Formel: C₉H₇BClNO₂
Lagertemperatur: Kühlschrank
MDL: MFCD22199342
CAS-Nummer: 1800483-72-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR