2-Phenyl-1,3-oxazole-4-boronic acid, pinacol ester 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-oxazole

OR300007-250MG
APOSOR300007-250MGEA 725 EUR
APOSOR300007-250MG
2-Phenyl-1,3-oxazole-4-boronic acid, pinacol ester 95%
2-Phenyl-1,3-oxazole-4-boronic acid, pinacol ester
Formel: C₁₅H₁₈BNO₃
Molecular Weight: 271,119 g/mol
Schmelzpunkt: 124…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18383302
CAS-Nummer: 885669-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR