Pyrido[3,4-b]pyrazine

Lieferant: Apollo Scientific
Achtung

Synonyme: pyridino[3,4-b]pyrazine

OR300013-1G
APOSOR300013-1GEA 313 EUR
APOSOR300013-1G
Pyrido[3,4-b]pyrazine
Pyrido[3,4-b]pyrazine
Formel: C₇H₅N₃
Molecular Weight: 131,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01646167
CAS-Nummer: 254-86-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR