3-Bromo-8-chloroimidazo[1,2-a]pyrazine

Lieferant: Apollo Scientific

OR300016-1G
APOSOR300016-1GEA 69.6 EUR
APOSOR300016-1G
3-Bromo-8-chloroimidazo[1,2-a]pyrazine
3-Bromo-8-chloroimidazo[1,2-a]pyrazine
Formel: C6H3BrClN3
Lagertemperatur: Raumtemperatur
MDL: MFCD09909653
CAS-Nummer: 143591-61-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR