6-Bromoimidazo[1,2-a]pyrazin-8-amine
Lieferant: Apollo Scientific
Synonyme:
8-Amino-6-bromoimidazo[1,2-a]pyrazine
OR300017-1G
APOSOR300017-1GEA
112
EUR
APOSOR300017-1G
6-Bromoimidazo[1,2-a]pyrazin-8-amine
6-Bromoimidazo[1,2-a]pyrazin-8-amine
Formel:
C₆H₅BrN₄ Molecular Weight: 213,0400 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00874115 CAS-Nummer: 117718-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...