6-Bromo-8-methoxyimidazo[1,2-a]pyrazine

Lieferant: Apollo Scientific

OR300022-1G
APOSOR300022-1GEA 324 EUR
APOSOR300022-1G
6-Bromo-8-methoxyimidazo[1,2-a]pyrazine
6-Bromo-8-methoxyimidazo[1,2-a]pyrazine
Formel: C₇H₆BrN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD09835036
CAS-Nummer: 63744-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR