3-Bromo-5-chloroimidazo[1,2-a]pyrazine

Lieferant: Apollo Scientific
Achtung

OR300023-1G
APOSOR300023-1GEA 1430 EUR
APOSOR300023-1G
3-Bromo-5-chloroimidazo[1,2-a]pyrazine
3-Bromo-5-chloroimidazo[1,2-a]pyrazine
Formel: C₆H₃BrClN₃
Molecular Weight: 232,465 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16995965
CAS-Nummer: 1243389-43-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR