8-Chloro[1,2,4]triazolo[4,3-a]pyrazine

Lieferant: Apollo Scientific

OR300026-1G
APOSOR300026-1GEA 55.1 EUR
APOSOR300026-1G
8-Chloro[1,2,4]triazolo[4,3-a]pyrazine
8-Chloro[1,2,4]triazolo[4,3-a]pyrazine
Formel: C₅H₃ClN₄
Molecular Weight: 154,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08272086
CAS-Nummer: 68774-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR