6-Chloro[1,2,4]triazolo[4,3-a]pyrazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-[1,2,4]triazolo[4,3-a]pyrazine

OR300027-1G
APOSOR300027-1GEA 382 EUR
APOSOR300027-1G
6-Chloro[1,2,4]triazolo[4,3-a]pyrazine
6-Chloro[1,2,4]triazolo[4,3-a]pyrazine
Formel: C₅H₃ClN₄
Molecular Weight: 154,557 g/mol
Schmelzpunkt: 195…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11043990
CAS-Nummer: 1314974-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR