6-Bromo-8-chloro-[1,2,4]triazolo[4,3-a]pyrazine
Lieferant: Apollo Scientific
Synonyme:
6-Bromo-8-chlorotriazolo[4,3-a]pyrazine
OR300030-1G
APOSOR300030-1GEA
1050
EUR
APOSOR300030-1G
6-Bromo-8-chloro-[1,2,4]triazolo[4,3-a]pyrazine
6-Bromo-8-chloro-[1,2,4]triazolo[4,3-a]pyrazine
Formel:
C₅H₂BrClN₄ Molecular Weight: 233,453 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20501878 CAS-Nummer: 1334135-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...