8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyrazine
Lieferant: Apollo Scientific
Synonyme:
8-Chloro-3-methyl-[1,2,4]triazolo[4,3-a]pyrazine, 8-Chloro-3-methyltriazolo[4,3-a]pyrazine
OR300032-1G
APOSOR300032-1GEA
43.5
EUR
APOSOR300032-1G
8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyrazine
8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyrazine
Formel:
C₆H₅ClN₄ Molecular Weight: 168,584 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08272087 CAS-Nummer: 68774-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...