3-Phenoxypyridine 98%

Lieferant: Apollo Scientific

OR300041-1G
APOSOR300041-1GEA 102 EUR
APOSOR300041-1G
3-Phenoxypyridine 98%
3-Phenoxypyridine
Formel: C₁₁H₉NO
Molecular Weight: 171,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234704
CAS-Nummer: 2176-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR