5-Amino-4-methylisoquinoline
Lieferant: Apollo Scientific
|
Achtung
|
OR300048-1G
APOSOR300048-1GEA
604
EUR
APOSOR300048-1G
5-Amino-4-methylisoquinoline
5-Amino-4-methylisoquinoline
Formel:
C₁₀H₁₀N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12922847 CAS-Nummer: 194032-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...