5-(Hydroxymethyl)isoquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: Isoquinolin-5-ylmethanol, (Isoquinolin-5-yl)methanol

OR300050-1G
APOSOR300050-1GEA 299 EUR
APOSOR300050-1G
5-(Hydroxymethyl)isoquinoline
5-(Hydroxymethyl)isoquinoline
Formel: C₁₀H₉NO
Molecular Weight: 159.185 g/mol
Schmelzpunkt: 101…102 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03840599
CAS-Nummer: 76518-57-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR