1-(Isoquinolin-8-yl)ethanone 95%

Lieferant: Apollo Scientific

OR300057-1G
APOSOR300057-1GEA 335 EUR
APOSOR300057-1G
1-(Isoquinolin-8-yl)ethanone 95%
1-(Isoquinolin-8-yl)ethanone
Formel: C₁₁H₉NO
Molecular Weight: 171.195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10699238
CAS-Nummer: 1053655-98-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR