1-Chloroisoquinoline-6-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Chloro-2-azanaphthalene-6-carboxylic acid, 6-Carboxy-1-chloroisoquinoline

OR300061-250MG
APOSOR300061-250MGEA 94.4 EUR
APOSOR300061-250MG
1-Chloroisoquinoline-6-carboxylic acid
1-Chloroisoquinoline-6-carboxylic acid
Formel: C₁₀H₆ClNO₂
Molecular Weight: 207,613 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15526626
CAS-Nummer: 1256787-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR