1-Chloroisoquinoline-6-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Chloro-6-formyl-2-azanaphthalene, 1-Chloroisoquinoline-6-carbaldehyde, 1-Chloro-6-formylisoquinoline

OR300074-1G
APOSOR300074-1GEA 1100 EUR
APOSOR300074-1G
1-Chloroisoquinoline-6-carboxaldehyde
1-Chloroisoquinoline-6-carboxaldehyde
Formel: C₁₀H₆ClNO
Molecular Weight: 191,614 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD15526625
CAS-Nummer: 1211528-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR