3-Chloroisoquinoline-6-carboxaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-6-formyl-2-azanaphthalene, 3-Chloro-6-formylisoquinoline, 3-Chloroisoquinoline-6-carbaldehyde
OR300076-500MG
APOSOR300076-500MGEA
429
EUR
APOSOR300076-500MG
3-Chloroisoquinoline-6-carboxaldehyde
3-Chloroisoquinoline-6-carboxaldehyde
Formel:
C₁₀H₆ClNO Molecular Weight: 191,614 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22380179 CAS-Nummer: 1337880-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...