3-Chloroisoquinoline-7-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-7-formylisoquinoline, 3-Chloro-2-azanaphthalene-7-carboxaldehyde

OR300077-500MG
APOSOR300077-500MGEA 369 EUR
APOSOR300077-500MG
3-Chloroisoquinoline-7-carboxaldehyde
3-Chloroisoquinoline-7-carboxaldehyde
Formel: C₁₀H₆ClNO
Molecular Weight: 191.614 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22380180
CAS-Nummer: 1337879-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR