1-Hydroxyisoquinoline-7-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR300080-500MG
APOSOR300080-500MGEA 0 EUR
APOSOR300080-500MG
1-Hydroxyisoquinoline-7-carboxylic acid
1-Hydroxyisoquinoline-7-carboxylic acid
Formel: C₁₀H₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22375649
CAS-Nummer: 1301214-62-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR