3-Bromo-6-chloroisoquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-6-chloro-2-azanaphthalene

OR300092-1G OR300092-250MG
APOSOR300092-1GEA 810 EUR
APOSOR300092-1G APOSOR300092-250MG
3-Bromo-6-chloroisoquinoline
3-Bromo-6-chloroisoquinoline
Formel: C₉H₅BrClN
Molecular Weight: 242.5 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18793407
CAS-Nummer: 1276056-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR