Methyl 5-bromoisoquinoline-1-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-1-(methoxycarbonyl)-2-azanaphthalene, 5-Bromo-1-(methoxycarbonyl)isoquinoline

OR300109-250MG OR300109-100MG
APOSOR300109-250MGEA 638 EUR
APOSOR300109-250MG APOSOR300109-100MG
Methyl 5-bromoisoquinoline-1-carboxylate
Methyl 5-bromoisoquinoline-1-carboxylate
Formel: C₁₁H₈BrNO₂
Molecular Weight: 266.091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19981746
CAS-Nummer: 1330750-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR