1-Bromo-7-chloroisoquinoline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-7-chloro-2-azanaphthalene
OR300112-250MG
APOSOR300112-250MGEA
81.2
EUR
APOSOR300112-250MG
1-Bromo-7-chloroisoquinoline
1-Bromo-7-chloroisoquinoline
Formel:
C₉H₅BrClN Molecular Weight: 242,5 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13190245 CAS-Nummer: 1196155-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...