6-Amino-3-chloroisoquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloroisoquinolin-6-amine, 6-Amino-3-chloro-2-azanaphthalene

OR300114-1G
APOSOR300114-1GEA 930 EUR
APOSOR300114-1G
6-Amino-3-chloroisoquinoline
6-Amino-3-chloroisoquinoline
Formel: C₉H₇ClN₂
Molecular Weight: 178,618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22380488
CAS-Nummer: 1374652-51-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR