1,6-Dichlorisochinolin 97%

Lieferant: Apollo Scientific

OR300125-5G
APOSOR300125-5GEA 71.2 EUR
APOSOR300125-5G
1,6-Dichlorisochinolin 97%
1,6-Dichloroisoquinoline
Formel: C₉H₅Cl₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD06738660
CAS-Nummer: 630421-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR