7-Chloro-1-iodoisoquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Chloro-1-iodo-2-azanaphthalene

OR300131-250MG
APOSOR300131-250MGEA 178 EUR
APOSOR300131-250MG
7-Chloro-1-iodoisoquinoline
7-Chloro-1-iodoisoquinoline
Formel: C₉H₅ClIN
Molecular Weight: 289,5 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14702501
CAS-Nummer: 1203579-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR