7-Bromoisoquinoline-1,3(2H,4H)-dione

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromo-1,3-dioxo-1,2,3,4-tetrahydroisoquinoline

OR300133-1G
APOSOR300133-1GEA 140 EUR
APOSOR300133-1G
7-Bromoisoquinoline-1,3(2H,4H)-dione
7-Bromoisoquinoline-1,3(2H,4H)-dione
Formel: C₉H₆BrNO₂
Molecular Weight: 240,053 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12756049
CAS-Nummer: 1033330-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR