7-Bromo-1,3-dichloroisoquinoline 70%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromo-1,3-dichloro-2-azanaphthalene

OR300137-100MG
APOSOR300137-100MGEA 31.9 EUR
APOSOR300137-100MG
7-Bromo-1,3-dichloroisoquinoline 70%
7-Bromo-1,3-dichloroisoquinoline
Formel: C₉H₄BrCl₂N
Molecular Weight: 276,945 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11112092
CAS-Nummer: 924271-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR