Methyl 1-methyl-1H-indazole-3-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR300142-1G
OR300142-250MG
APOSOR300142-1GEA
34.8
EUR
APOSOR300142-1G
APOSOR300142-250MG
Methyl 1-methyl-1H-indazole-3-carboxylate
Methyl 1-methyl-1H-indazole-3-carboxylate
Formel:
C₁₀H₁₀N₂O₂ Molecular Weight: 190.199 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03840643 CAS-Nummer: 109216-60-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...