Methyl 1-methyl-1H-indazole-3-carboxylate

Lieferant: Apollo Scientific
Achtung

OR300142-1G OR300142-250MG
APOSOR300142-1GEA 34.8 EUR
APOSOR300142-1G APOSOR300142-250MG
Methyl 1-methyl-1H-indazole-3-carboxylate
Methyl 1-methyl-1H-indazole-3-carboxylate
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190.199 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03840643
CAS-Nummer: 109216-60-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR