3-Methoxyisoquinoline-7-boronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Borono-3-methoxyisoquinoline, 7-Borono-3-methoxy-2-azanaphthalene
OR300146-100MG
APOSOR300146-100MGEA
145
EUR
APOSOR300146-100MG
3-Methoxyisoquinoline-7-boronic acid
3-Methoxyisoquinoline-7-boronic acid
Formel:
C₁₀H₁₀BNO₃ Molecular Weight: 203,002 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD19982319 CAS-Nummer: 1645213-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...