3-Chloroisoquinoline-5-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Borono-3-chloro-2-azanaphthalene, 5-Borono-3-chloroisoquinoline, (3-Chloroisoquinolin-5-yl)boronic acid

OR300151-500MG OR300151-100MG
APOSOR300151-500MGEA 435 EUR
APOSOR300151-500MG APOSOR300151-100MG
3-Chloroisoquinoline-5-boronic acid
3-Chloroisoquinoline-5-boronic acid
Formel: C₉H₇BClNO₂
Molecular Weight: 207.421 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22199341
CAS-Nummer: 1429665-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR