Ethyl 5-isopropyl-1-methyl-1H-pyrazole-3-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR300152-250MG
APOSOR300152-250MGEA
58
EUR
APOSOR300152-250MG
Ethyl 5-isopropyl-1-methyl-1H-pyrazole-3-carboxylate
Ethyl 5-isopropyl-1-methyl-1H-pyrazole-3-carboxylate
Formel:
C₁₀H₁₆N₂O₂ Molecular Weight: 196.246 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10699186 CAS-Nummer: 1170483-34-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...