Methyl 4-bromo-1H-indole-7-carboxylate 95+%

Lieferant: Apollo Scientific
Achtung

OR300169-250MG
APOSOR300169-250MGEA 241 EUR
APOSOR300169-250MG
Methyl 4-bromo-1H-indole-7-carboxylate 95+%
Methyl 4-bromo-1H-indole-7-carboxylate
Formel: C₁₀H₈BrNO₂
Molecular Weight: 254,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20487964
CAS-Nummer: 1224724-39-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR