2-(1-Benzyl-1H-pyrazol-5-yl)acetic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(1-Benzyl-1H-pyrazol-3-yl)acetic acid
OR300171-250MG
APOSOR300171-250MGEA
221
EUR
APOSOR300171-250MG
2-(1-Benzyl-1H-pyrazol-5-yl)acetic acid
2-(1-Benzyl-1H-pyrazol-5-yl)acetic acid
Formel:
C₁₂H₁₂N₂O₂ Molecular Weight: 216,24 g/mol |
MDL:
MFCD28530874 CAS-Nummer: 1194803-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...