N-Methylbicyclo[1.1.1]pentan-1-amine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR300175-250MG
APOSOR300175-250MGEA 404 EUR
APOSOR300175-250MG
N-Methylbicyclo[1.1.1]pentan-1-amine hydrochloride
N-Methylbicyclo[1.1.1]pentan-1-amine hydrochloride
Formel: C₆H₁₁N·ClH
Molecular Weight: 133,619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31417451
CAS-Nummer: 1882089-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR