N-Benzylbicyclo[1.1.1]pentan-1-amine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR300176-250MG
APOSOR300176-250MGEA 638 EUR
APOSOR300176-250MG
N-Benzylbicyclo[1.1.1]pentan-1-amine hydrochloride
N-Benzylbicyclo[1.1.1]pentan-1-amine hydrochloride
Formel: C₁₂H₁₅N·ClH
Molecular Weight: 209,715 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31536714
CAS-Nummer: 2187435-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR