N-Ethylbicyclo[1.1.1]pentan-1-amine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR300180-250MG
APOSOR300180-250MGEA 580 EUR
APOSOR300180-250MG
N-Ethylbicyclo[1.1.1]pentan-1-amine hydrochloride
N-Ethylbicyclo[1.1.1]pentan-1-amine hydrochloride
Formel: C₇H₁₃N·ClH
Molecular Weight: 147,646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31536718
CAS-Nummer: 2187435-35-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR